Learn More
2B4/CD244/SLAMF4 Antibody (244F4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-00223-0.025MG
This item is not returnable.
View return policy
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| 2B4/CD244/SLAMF4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000 | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| Rat | |
| Affinity Purified | |
| RUO | |
| 51744 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry | |
| 244F4 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD244 | |
| The immunogen for this antibody is CD244. | |
| 0.025 mg | |
| Primary | |
| Mouse | |
| IgG2a |