missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (244F4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 197.00 - € 622.00
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
|---|---|
| Clone | 244F4 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000 |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18096666
|
Novus Biologicals
NBP2-00223-100UG |
100 μg |
€ 486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047246
|
Novus Biologicals
NBP2-00223-500UG |
500 μg |
€ 622.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18001867
|
Novus Biologicals
NBP2-00223-0.025MG |
0.025 mg |
€ 197.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| 2B4/CD244/SLAMF4 | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 51744 | |
| The immunogen for this antibody is CD244. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 244F4 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| CD244 | |
| IgG2a | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title