Learn More
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
Product Code. p-7029812
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15487675
Brand: Thermo Scientific Alfa Aesar H27321.MD
Request Bulk or Custom Format
CAS: 61478-28-2 | C34H37NO2P2 | 553.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61478-28-2
Molecular Formula C34H37NO2P2
Molecular Weight (g/mol) 553.62
MDL Number MFCD01631275
InChI Key BFMKBYZEJOQYIM-FEHSQGEHNA-N
Synonym 2s,4s---n-boc-4-diphenylphosphino-2-diphenylphosphinomethyl-pyrrolidine, bppm, s,s-bppm, 2s,4s-1-tert-butoxycarbonyl-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---2-diphenylphosphinomethyl-4-diphenylphosphino-n-t-butoxycarbonyl pyrrolidine, 2s,4s-tert-butyl 4-diphenylphosphino-2-diphenylphosphino methyl pyrrolidine-1-carboxylate, tert-butyl 2s,4s-4-diphenylphosphanyl-2-diphenylphosphanyl methyl pyrrolidine-1-carboxylate, 2s,4s-1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine
PubChem CID 10209545
IUPAC Name tert-butyl (2S,4S)-4-diphenylphosphanyl-2-(diphenylphosphanylmethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 100°C to 103°C
Quantity 250 mg
Formula Weight 553.62
Chemical Name or Material (2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine