Learn More
3-(2-Furyl)acrolein, 99%
Product Code. 11449973
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449973 5 g 5g
11459973 25 g 25g
11469973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11449973 Supplier Thermo Scientific Alfa Aesar Supplier No. A13042.06
Request Bulk or Custom Format
CAS: 623-30-3 | C7H6O2 | 122.123 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 623-30-3
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00003256
InChI Key VZIRCHXYMBFNFD-HNQUOIGGSA-N
Synonym 3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein
PubChem CID 1549521
IUPAC Name (E)-3-(furan-2-yl)prop-2-enal
SMILES C1=COC(=C1)C=CC=O
Specifications
Melting Point 49°C to 52°C
Boiling Point 95°C (5 mmHg)
Flash Point 99°C (210°F)
Quantity 5 g
UN Number UN1759
Beilstein 107570
Sensitivity Air sensitive
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material 3-(2-Furyl)acrolein
compliance-icons
Product Identifier
  • trans-beta-(2-Furylacrolein)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3-(2-furanyl)-2-propenal