Learn More
3,3'-Dimethoxybenzil, 99+%
Product Code. 11352497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352497 5 g 5g
11342497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11352497 Supplier Thermo Scientific Alfa Aesar Supplier No. L11564.06
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 40101-17-5 | C16H14O4 | 270.284 g/mol
3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40101-17-5
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00038221
InChI Key PJGXOGKIVAJFTE-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzil, 1,2-bis 3-methoxyphenyl ethane-1,2-dione, ethanedione, bis 3-methoxyphenyl, 1,2-ethanedione, 1,2-bis 3-methoxyphenyl, bis 3-methoxyphenyl ethane-1,2-dione, bis 3-methoxyphenyl ethanedione, acmc-20a2cz, 3,3'-dimethoxy benzil, d01vau, benzil-based compound, 22
PubChem CID 123493
IUPAC Name 1,2-bis(3-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=CC(=C1)C(=O)C(=O)C2=CC(=CC=C2)OC
Specifications
Melting Point 81°C to 83°C
Quantity 5 g
Beilstein 2382124
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity ≥99%
Chemical Name or Material 3,3'-Dimethoxybenzil