missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzonitrile, 97%

Product Code. 11412827
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412827 1 g 1g
11422827 5 g 5g
11432827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412827 Supplier Thermo Scientific Alfa Aesar Supplier No. A15460.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 27126-93-8 | C9H3F6N | 239.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27126-93-8
Molecular Formula C9H3F6N
Molecular Weight (g/mol) 239.12
MDL Number MFCD00000379
InChI Key CZKHHAOIHXHOSR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
PubChem CID 96746
IUPAC Name 3,5-bis(trifluoromethyl)benzonitrile
SMILES FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F

Specifications

Melting Point 16°C to 20°C
Density 1.421
Boiling Point 154°C to 156°C
Flash Point 72°C (161°F)
Refractive Index 1.4175
Quantity 1 g
UN Number UN3276
Beilstein 3552650
Formula Weight 239.12
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.