missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamido-3-deoxy-D-glucose, TRC

Product Code. 30179099
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179099 50 mg 50mg
30179249 25 mg 25mg
30179629 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179099 Supplier Toronto Research Chemicals Supplier No. TRCA15800050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14086-88-5
Molecular Formula C8 H15 N O6
Molecular Weight (g/mol) 221.21
IUPAC Name N-[(3R,4S,5S,6R)-2,3,5-trihydroxy-6-(hydroxymethyl)oxan-4-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)C(O)O[C@H](CO)[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H15NO6/c1-3(11)9-5-6(12)4(2-10)15-8(14)7(5)13/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5+,6-,7-,8?/m1/s1
Formula Weight 221.0899
Chemical Name or Material 3-Acetamido-3-deoxy-D-glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.