missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromopyridine, 97%

Product Code. p-7047847
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11339825

Brand: Thermo Scientific Alfa Aesar L19839.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 39856-58-1 | C5H5BrN2 | 173.013 g/mol

Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39856-58-1
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD00234064
InChI Key HKDVVTLISGIPFE-UHFFFAOYSA-N
Synonym 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine
PubChem CID 642816
IUPAC Name 2-bromopyridin-3-amine
SMILES C1=CC(=C(N=C1)Br)N

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
Beilstein 109830
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Amino-2-bromopyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.