missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1H-indazole, 97%, Thermo Scientific™

Product Code. p-7032111
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15466267

Brand: Thermo Scientific Alfa Aesar H33460.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Microwave heating of 2-halobenzaldehydes or 2-haloacetophenones with phenylhydrazines at 160°C for 10 min quantitatively yielded the arylhydrazones, which were further cyclized to give 1-aryl-1H-indazoles via CuI/diamine-catalyzed N-arylation under microwave heating (160°C, 10 min). Good to excellent yields were observed for 2-iodo, 2-bromo, and 2-chloro benzaldehydes or acetophenones.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate 2-7°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914311-50-5
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09909580
InChI Key IXXXOIXGQFPLIH-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine
PubChem CID 11975123
IUPAC Name 4-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(Br)=C12

Specifications

Melting Point 142°C to 144°C
Quantity 250 mg
Solubility Information Slightly soluble.
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromo-1H-indazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.