missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 3-Bromocytisine

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent agonist of α4β4, α4β2 and α7 nACh receptors (IC50 values are 0.28, 0.30 and 31.6 nM respectively). Displays different effects on high (HS) and low (LS) ACh sensitivity α4β2 nAChRs (EC50 values are 0.008 and 0.05 μM respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 269.14 |
| Purity | >98% |
| Molecular Weight (g/mol) | 269.142 |
| Chemical Name or Material | 3-Bromocytisine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction