missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine hydrochloride, 99%, Thermo Scientific™

Product Code. 11309786 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 11309786

Brand: Thermo Scientific Acros 174320050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 64051-79-2
Molecular Formula C5H11NO·ClH
Molecular Weight (g/mol) 137.61
InChI Key VLECDMDGMKPUSK-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine hydrochloride, piperidin-3-ol hydrochloride, 3-hydroxypiperidine hcl, 3-piperidinol hydrochloride, piperidine-3-ol hydrochloride, 3-hydroxy-piperidine hydrochloride, 3-piperidinol, hydrochloride, piperidine-3-ol hcl, 3-hydroxypiperidine.hcl, 3-hydroxypiperidinehydrochloride
PubChem CID 2723962
IUPAC Name piperidin-3-ol;hydrochloride
SMILES C1CC(CNC1)O.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 196.6°C to 198.7°C
Color White
Elemental Analysis 42.97 to 44.28% C, 9.90 to 10.33% N
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 5 g
Formula Weight 137.61
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Hydroxypiperidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxypiperidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.