missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-2-methylpropanoic Acid, TRC

Product Code. 30483779
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483779 1 g 1g
30471340 5 g 5g
30475431 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483779 Supplier Toronto Research Chemicals Supplier No. TRCM2539501G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26473-47-2
Molecular Formula C4 H8 O2 S
Molecular Weight (g/mol) 120.17
Synonym (2RS)-2-Methyl-3-sulfanyl-propanoic Acid, Captopril Imp. C (EP), Isobutyric acid, β-mercapto- (4CI), Propionic acid, 3-mercapto-2-methyl- (8CI), 3-Mercapto-2-methylpropanoic acid, 3-Mercapto-2-methylpropionic acid, 3-Mercaptoisobutyric acid
IUPAC Name 2-methyl-3-sulfanylpropanoic acid
SMILES CC(CS)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H8O2S/c1-3(2-7)4(5)6/h3,7H,2H2,1H3,(H,5,6)
Formula Weight 120.0245
Percent Purity >95%
Chemical Name or Material (2RS)-2-Methyl-3-sulfanyl-propanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.