CAS RN 26473-47-2
IUPAC Name:
sodium 2-methyl-3-sulfanylpropanoate
Synonyms:
sodium 2-methyl-3-sulfanylpropanoate
Molecular Weight (g/mol):
142.15
Molecular Formula:
C4H7NaO2S
InChi Key:
FOTPRJUNQGWLCM-UHFFFAOYNA-M
SMILES:
[Na+].CC(CS)C([O-])=O
1
–
2
of
2
results
1
(2RS)-2-Methyl-3-sulfanyl-propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Mercapto-2-methylpropanoic Acid, TRC
CAS: 26473-47-2 Molecular Formula: C4 H8 O2 S Molecular Weight (g/mol): 120.17 Synonym: (2RS)-2-Methyl-3-sulfanyl-propanoic Acid,Captopril Imp. C (EP),Isobutyric acid, β-mercapto- (4CI),Propionic acid, 3-mercapto-2-methyl- (8CI),3-Mercapto-2-methylpropanoic acid,3-Mercapto-2-methylpropionic acid,3-Mercaptoisobutyric acid IUPAC Name: 2-methyl-3-sulfanylpropanoic acid SMILES: CC(CS)C(=O)O