Learn More
3-Methoxyphenylhydrazine hydrochloride, 98%
Product Code. 11327985
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11327985
Brand: Thermo Scientific Alfa Aesar L08091.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 39232-91-2 | C7H11ClN2O | 174.628 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39232-91-2
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
MDL Number MFCD00044416
InChI Key GMXFZBZOVZOYNQ-UHFFFAOYSA-N
Synonym 3-methoxyphenylhydrazine hydrochloride, 3-methoxyphenyl hydrazine hydrochloride, hydrazine, 3-methoxyphenyl-, monohydrochloride, 3-methoxy-phenylhydrazine hydrochloride, acmc-20an1f, ksc491e7n, 3-methoxyphenyl-hydrazine hcl, 3-methoxyphenylhydrazine, hcl, 3-methoxyphenylhydrazin hydrochloride, m-methoxyphenylhydrazine hydrochloride
PubChem CID 2723940
IUPAC Name (3-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=CC(=C1)NN.Cl
Specifications
Melting Point ∽142°C (decomposition)
Quantity 1 g
Beilstein 5304389
Formula Weight 174.63
Percent Purity 98%
Chemical Name or Material 3-Methoxyphenylhydrazine hydrochloride