missing translation for 'onlineSavingsMsg'
Learn More

3-O-Alpha-D-Glucosyl Isomaltol, TRC

Product Code. 30004184
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004184 10 mg 10mg
30004724 2.5 mg 2.5mg
30006338 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004184 Supplier Toronto Research Chemicals Supplier No. TRCG59687510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85559-61-1
Molecular Formula C12H16O8
Molecular Weight (g/mol) 288.25
Synonym 1-[3-(α-D-Glucopyranosyloxy)-2-furanyl]ethanone, 2-Acetylfuran-3yl-α-D-glucopyranoside;
IUPAC Name 1-[3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyfuran-2-yl]ethanone
SMILES O=C(C)C(OC=C1)=C1O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H16O8/c1-5(14)11-6(2-3-18-11)19-12-10(17)9(16)8(15)7(4-13)20-12/h2-3,7-10,12-13,15-17H,4H2,1H3/t7-,8-,9+,10-,12+/m1/s1
Formula Weight 288.0845
Percent Purity >95%
Chemical Name or Material 3-O-alpha-D-Glucosyl Isomaltol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.