missing translation for 'onlineSavingsMsg'
Learn More

7-Pivalamido Cefazolin, TRC

Product Code. 30352915
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352915 5 mg 5mg
30350512 10 mg -
30361527 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352915 Supplier Toronto Research Chemicals Supplier No. TRCC2341085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2384108-14-7
Molecular Formula C16H20N4O4S3
Molecular Weight (g/mol) 428.55
Synonym Cefazolin EP Impurity B, (6R,7R)-3-(((5-Methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-7-pivalamido-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
IUPAC Name (6R,7R)-7-(2,2-dimethylpropanoylamino)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)C(C)(C)C)C3=O)C(=O)O)s1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H20N4O4S3/c1-7-18-19-15(27-7)26-6-8-5-25-12-9(17-14(24)16(2,3)4)11(21)20(12)10(8)13(22)23/h9,12H,5-6H2,1-4H3,(H,17,24)(H,22,23)/t9-,12-/m1/s1
Formula Weight 428.0647
Chemical Name or Material 7-Pivalamido Cefazolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.