missing translation for 'onlineSavingsMsg'
Learn More

N-(Deoxyguanosin-8-yl)-2-amino-1-methyl-6-phenylimidazo[4,5-Beta]pyridine, TRC

Product Code. 30352994
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.50mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352994 5 mg 5mg
30358193 1 mg 1mg
30364716 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352994 Supplier Toronto Research Chemicals Supplier No. TRCD2396305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142784-25-6
Molecular Formula C23 H23 N9 O4
Molecular Weight (g/mol) 489.49
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-imino-8-[(1-methyl-6-phenylimidazo[4,5-b]pyridin-2-yl)amino]-3H-purin-6-one
SMILES Cn1c(Nc2nc3C(=O)NC(=N)Nc3n2[C@H]4C[C@H](O)[C@@H](CO)O4)nc5ncc(cc15)c6ccccc6

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H23N9O4/c1-31-13-7-12(11-5-3-2-4-6-11)9-25-18(13)27-22(31)30-23-26-17-19(28-21(24)29-20(17)35)32(23)16-8-14(34)15(10-33)36-16/h2-7,9,14-16,33-34H,8,10H2,1H3,(H3,24,28,29,35)(H,25,26,27,30)/t14-,15+,16+/m0/s1
Formula Weight 489.1873
Chemical Name or Material N-(Deoxyguanosin-8-yl)-2-amino-1-methyl-6-phenylimidazo[4,5-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.