missing translation for 'onlineSavingsMsg'
Learn More

Cefamandole Lithium, TRC

Product Code. 30353143
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353143 100 mg 100mg
30351434 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30353143 Supplier Toronto Research Chemicals Supplier No. TRCC237495100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58648-57-0
Molecular Formula C18 H17 N6 O5 S2 . Li
Molecular Weight (g/mol) 468.44
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(hydroxyphenylacetyl)amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, monolithium salt, [6R-[6α,7β(R*)]]- (9CI), Cefamandole lithium, Cefamandole lithium salt
IUPAC Name lithium;(6R,7R)-7-[[(2R)-2-hydroxy-2-phenylacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Li+].Cn1nnnc1SCC2=C(N3[C@H](SC2)[C@H](NC(=O)[C@H](O)c4ccccc4)C3=O)C(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H18N6O5S2.Li/c1-23-18(20-21-22-23)31-8-10-7-30-16-11(15(27)24(16)12(10)17(28)29)19-14(26)13(25)9-5-3-2-4-6-9;/h2-6,11,13,16,25H,7-8H2,1H3,(H,19,26)(H,28,29);/q;+1/p-1/t11-,13-,16-;/m1./s1
Formula Weight 468.0862
Chemical Name or Material Cefamandole Lithium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.