missing translation for 'onlineSavingsMsg'
Learn More

Conduritol B, TRC

Product Code. 30354402
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354402 5 mg
30363921 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354402 Supplier Toronto Research Chemicals Supplier No. TRCC6650005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25348-64-5
Molecular Formula C6 H10 O4
Molecular Weight (g/mol) 146.14
IUPAC Name (1R,2S,3S,4R)-cyclohex-5-ene-1,2,3,4-tetrol
SMILES O[C@@H]1C=C[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H10O4/c7-3-1-2-4(8)6(10)5(3)9/h1-10H/t3-,4-,5+,6+/m1/s1
Formula Weight 146.0579
Chemical Name or Material Conduritol B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.