missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Vindoline, TRC

Product Code. 30358776
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358776 100 mg 100mg
30365792 25 mg 25mg
30354288 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358776 Supplier Toronto Research Chemicals Supplier No. TRCD288745100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3633-92-9
Molecular Formula C23H30N2O5
Molecular Weight (g/mol) 414.49
Synonym (2β,3β,4β,5α,12R,19α)-6,7-Didehydro-3,4-dihydroxy-16-methoxy-1-methyl-aspidospermidine-3-carboxylic Acid Methyl Ester, 4-Desacetylvindoline, Deacetylvindoline, NSC 91993
IUPAC Name methyl (3aR,3a1R,4R,5S,5aR,10bR)-3a-ethyl-4,5-dihydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate
SMILES [H][C@]12[C@](C=CCN2CC3)(CC)[C@@H](O)[C@@](C(OC)=O)(O)[C@@]4([H])[C@]13C5=CC=C(OC)C=C5N4C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H30N2O5/c1-5-21-9-6-11-25-12-10-22(17(21)25)15-8-7-14(29-3)13-16(15)24(2)18(22)23(28,19(21)26)20(27)30-4/h6-9,13,17-19,26,28H,5,10-12H2,1-4H3/t17-,18+,19+,21+,22+,23-/m0/s1
Formula Weight 414.22
Chemical Name or Material Desacetyl Vindoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.