missing translation for 'onlineSavingsMsg'
Learn More

Lisadimate (>85%), TRC

Product Code. 30478837
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478837 100 mg 100mg
30471666 10 mg 10mg
30466960 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478837 Supplier Toronto Research Chemicals Supplier No. TRCL460000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-44-7
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.21
Synonym Glyceryl p-aminobenzoate, Escalol 106, Glycerol, 1-(p-aminobenzoate), 2,3-Dihydroxypropyl 4-aminobenzoate, 4-Aminobenzoic acid 1-glyceryl ester, NIPA GMPA
IUPAC Name 2,3-dihydroxypropyl 4-aminobenzoate
SMILES Nc1ccc(cc1)C(=O)OCC(O)CO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H13NO4/c11-8-3-1-7(2-4-8)10(14)15-6-9(13)5-12/h1-4,9,12-13H,5-6,11H2
Formula Weight 211.0845
Chemical Name or Material Lisadimate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.