missing translation for 'onlineSavingsMsg'
Learn More

5'-O-Methylthymidine, TRC

Product Code. 30484005
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484005 5 mg 5mg
30467819 25 mg 25mg
30475809 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484005 Supplier Toronto Research Chemicals Supplier No. TRCM3399585MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14504-60-0
Molecular Formula C11H16N2O5
Molecular Weight (g/mol) 256.26
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(methoxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES COC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C)C(=O)NC2=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H16N2O5/c1-6-4-13(11(16)12-10(6)15)9-3-7(14)8(18-9)5-17-2/h4,7-9,14H,3,5H2,1-2H3,(H,12,15,16)/t7-,8+,9+/m0/s1
Formula Weight 256.1059
Chemical Name or Material 5'-O-Methylthymidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.