missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-[4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]phenyl]-1-butanone, TRC

Product Code. 17990975
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990975 1 g 1g
17973845 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17990975 Supplier Toronto Research Chemicals Supplier No. TRCC3785151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67987-08-0
Molecular Formula C32 H36 Cl2 N2 O3
Molecular Weight (g/mol) 567.55
Synonym 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-[4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]phenyl]-, Haloperidol Related Compound A (USP)
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-[4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]phenyl]butan-1-one
SMILES OC1(CCN(CCCC(=O)c2ccc(cc2)N3CCC(O)(CC3)c4ccc(Cl)cc4)CC1)c5ccc(Cl)cc5

Specifications

Quantity 1 g
InChI Formula InChI=1S/C32H36Cl2N2O3/c33-27-9-5-25(6-10-27)31(38)15-20-35(21-16-31)19-1-2-30(37)24-3-13-29(14-4-24)36-22-17-32(39,18-23-36)26-7-11-28(34)12-8-26/h3-14,38-39H,1-2,15-23H2
Formula Weight 566.2103
Percent Purity >95%
Chemical Name or Material 4,4'-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]-butyrophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.