missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)butyric acid, 98%, Thermo Scientific™

Product Code. 11415570
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415570 5 g 5g
11405570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415570 Supplier Thermo Scientific Alfa Aesar Supplier No. H25839.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149437-76-3
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD00667222
InChI Key ZBQROUOOMAMCQW-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl butyric acid, 5-4-fluorophenyl-5-oxopentanoic acid, 4-4-fluorobenzoyl butyricacid, 5-4-fluorophenyl-5-oxovaleric acid, 4-fluorobenzoylbutyric acid, 5-4'-fluorophenyl-5-oxopentanoic acid, 4-4-fluorobenzoyl butanoic acid, 4-4-fluorobenzoyl-butyric acid, 4-fluorophenyl-5-oxopentanoic acid, 4-4'-fluorobenzoyl butyric acid
PubChem CID 689096
IUPAC Name 5-(4-fluorophenyl)-5-oxopentanoic acid
SMILES C1=CC(=CC=C1C(=O)CCCC(=O)O)F

Specifications

Melting Point 141°C to 144°C
Quantity 5 g
Formula Weight 210.21
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorobenzoyl)butyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.