missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde, 90%, Thermo Scientific™

Product Code. 10346234
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346234 1 g Amber Glass Bottle 1g
11396775 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346234 Supplier Thermo Scientific Maybridge Supplier No. CC53004DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 871252-64-1
Molecular Formula C13H8F3NO
Molecular Weight (g/mol) 251.208
MDL Number MFCD06797372
InChI Key SBXWMOPSHGIWEN-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
PubChem CID 2763359
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 251.21
Percent Purity 90%
Chemical Name or Material 4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.