missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzonitrile, 98+%

Product Code. 10163990 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10163990

Brand: Thermo Scientific Acros 167580250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1443-80-7 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1443-80-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001825
InChI Key NLPHXWGWBKZSJC-UHFFFAOYSA-N
Synonym 4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile
PubChem CID 74044
IUPAC Name 4-acetylbenzonitrile
SMILES CC(=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 55°C to 58°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4CN
Quantity 25 g
Beilstein 10, 695
Formula Weight 145.16
Percent Purity 98+%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 4-Acetylbenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylbenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.