missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzaldehyde, 98%

Product Code. 11407746
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11407746

Brand: Thermo Scientific Alfa Aesar A14716.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2426-87-1 | C15H14O3 | 242.27 g/mol

4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N?-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

Solubility
Soluble in chloroform, methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-87-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003365
InChI Key JSHLOPGSDZTEGQ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
PubChem CID 75506
IUPAC Name 3-methoxy-4-phenylmethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 61°C to 65°C
Boiling Point 213°C to 215°C (3.5 mmHg)
Quantity 100 g
Beilstein 1464258
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,methanol.
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.