Learn More
4'-Bromo-3'-nitroacetophenone 99+%, ACROS Organics™
Product Code. 10705981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. 10705981
Brand: Thermo Scientific Acros 136770050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 18640-58-9
CAS Min % 99.0
CAS Max % 100.0
Melting Point 118.0°C to 120.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H6BrNO3
Linear Formula Br(NO2)C6H3COCH3
MDL Number MFCD00016985
Quantity 5 g
Synonym 1-4-bromo-3-nitrophenyl ethanone, 4'-bromo-3'-nitroacetophenone, 1-4-bromo-3-nitrophenyl ethan-1-one, 4-bromo-3-nitroacetophenone, 4-bromo-3'-nitroacetophenone, ethanone, 1-4-bromo-3-nitrophenyl, acmc-209eny, 3-nitro-4-bromoacetophenone, 4-bromo-3-nitro acetophenone, 3'-nitro-4'-bromoacetophenone
InChI Key YFVOFFKNHQTQQE-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
IUPAC Name 1-(4-bromo-3-nitrophenyl)ethan-1-one
Molecular Weight (g/mol) 244.04
PubChem CID 87737
Formula Weight 244.04
Percent Purity ≥99%
Physical Form Amorphous Powder
Chemical Name or Material 4'-Bromo-3'-nitroacetophenone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Bromo-3'-nitroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.