missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylthieno[2,3-d]pyrimidine, TRC

Product Code. 30351048
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351048 100 mg -
30362059 50 mg 50mg
30359142 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351048 Supplier Toronto Research Chemicals Supplier No. TRCC368935100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56843-79-9
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.65
IUPAC Name 4-chloro-2-methylthieno[2,3-d]pyrimidine
SMILES Cc1nc(Cl)c2ccsc2n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H5ClN2S/c1-4-9-6(8)5-2-3-11-7(5)10-4/h2-3H,1H3
Formula Weight 183.9862
Chemical Name or Material 4-Chloro-2-methylthieno[2,3-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.