Learn More
4'-Chloro-N-methylformanilide, TRC
Product Code. 30353445
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353445 500 mg -
30354553 250 mg 250mg
30360321 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30353445 Supplier Toronto Research Chemicals Supplier No. TRCC428878500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26772-93-0
Molecular Formula C8H8NOCl
Molecular Weight (g/mol) 169.61
IUPAC Name N-(4-chlorophenyl)-N-methylformamide
SMILES CN(C=O)c1ccc(Cl)cc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H8ClNO/c1-10(6-11)8-4-2-7(9)3-5-8/h2-6H,1H3
Formula Weight 169.0294
Chemical Name or Material 4'-Chloro-N-methylformanilide