missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxynonenal Antibody (12F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59353-25ug
This item is not returnable.
View return policy
Description
4-Hydroxynonenal Monoclonal specifically detects 4-Hydroxynonenal in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| 4-Hydroxynonenal | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Specific for 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxynonenal. Does not cross-react with 4-Hydroxy-2-hexenal, Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins. | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 12F7 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| (2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4-hydroxy-trans-2-nonenal | |
| Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH) | |
| 25 μg | |
| Primary | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction