missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxynonenal Antibody (12F7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 197.00 - € 662.00
Specifications
| Antigen | 4-Hydroxynonenal |
|---|---|
| Clone | 12F7 |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216901
|
Novus Biologicals
NBP2-59353 |
100 μg |
€ 662.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633699
|
Novus Biologicals
NBP2-59353-25ug |
25 μg |
€ 197.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Hydroxynonenal Monoclonal specifically detects 4-Hydroxynonenal in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| 4-Hydroxynonenal | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| (2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4-hydroxy-trans-2-nonenal | |
| IgG1 | |
| Protein G purified |
| 12F7 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH) | |
| Primary | |
| Specific for 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxynonenal. Does not cross-react with 4-Hydroxy-2-hexenal, Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title