Learn More
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid, 97%, Thermo Scientific™
Product Code. 10374272
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10374272 1 g Amber Glass Bottle 1g
10140732 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10374272 Supplier Thermo Scientific Maybridge Supplier No. CC13501DA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 532391-89-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key XWYHWMDBIRZPFG-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carboxylic acid, 2h-1,4-benzoxazine-7-carboxylic acid, 3,4-dihydro-4-methyl, pubchem22096, 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxyl, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carboxylic acid
PubChem CID 2795296
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-7-carboxylic acid
SMILES CN1CCOC2=C1C=CC(=C2)C(=O)O
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid