CAS RN 532391-89-2
CAS RN 532391-89-2
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid, 97%, Thermo Scientific™
CAS: 532391-89-2 Molecular Formula: C10H11NO3 Molecular Weight (g/mol): 193.202 InChI Key: XWYHWMDBIRZPFG-UHFFFAOYSA-N Synonym: 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxylic acid,4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carboxylic acid,2h-1,4-benzoxazine-7-carboxylic acid, 3,4-dihydro-4-methyl,pubchem22096,4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxyl,4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carboxylic acid PubChem CID: 2795296 IUPAC Name: 4-methyl-2,3-dihydro-1,4-benzoxazine-7-carboxylic acid SMILES: CN1CCOC2=C1C=CC(=C2)C(=O)O