missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyridine-3-carboxylic acid, 97%

Product Code. p-4228830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10064257 1 g Glass bottle 1g
10413068 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10064257 Supplier Thermo Scientific Acros Supplier No. 429420010
Add to basket
This item is not returnable. View return policy

CAS: 3222-50-2 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-50-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00128869
InChI Key ZKUZSTXNVMIDCY-UHFFFAOYSA-N
Synonym 4-methylnicotinic acid, 4-methylpyridine-3-carboxylicacid, 4-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 4-methyl, 4-methyl-nicotinic acid, pubchem16270, acmc-1ags3, ksc223o4t, cbi-bb zero/009371, 3-pyridinecarboxylic acid,4-methyl
PubChem CID 229163
IUPAC Name 4-methylpyridine-3-carboxylic acid
SMILES CC1=C(C=NC=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 214.0°C to 215.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylpyridine-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylpyridine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.