Learn More
Schneider Gerd™ 4301 Stainless Steel Spoon
Brand:  Schneider Gerd™ 246
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10672835
  • € 11.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Specifications
1 Pk.
For Chemical
Stainless Steel (4301)
Spoon
35 x 48 mm
300 mm