Learn More
5-Benzyloxyindole-3-carboxaldehyde, 98%
Product Code. 11354464
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354464 5 g 5g
11364464 250 mg 250mg
11344464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11354464 Supplier Thermo Scientific Alfa Aesar Supplier No. L01117.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 6953-22-6 | C16H13NO2 | 251.285 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6953-22-6
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.285
MDL Number MFCD00014562
InChI Key DJGNUBADRQIDNQ-UHFFFAOYSA-N
Synonym 5-benzyloxyindole-3-carboxaldehyde, 5-benzyloxyindole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-phenylmethoxy-1h-indole-3-carbaldehyde, 5-benzyloxy-3-formylindole, 5-benzyloxy-indole-3-carboxaldehyde, 5-benzyloxy-3-formyl-1h-indole, 5-benzyloxy-3-indolecarboxaldehyde, 5-benzyloxy indole-3-carboxyaldehyde
PubChem CID 81398
IUPAC Name 5-phenylmethoxy-1H-indole-3-carbaldehyde
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3C=O
Specifications
Melting Point 239°C to 242°C
Quantity 5 g
Beilstein 208873
Sensitivity Air Sensitive
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 5-Benzyloxyindole-3-carboxaldehyde