Learn More
5-Chloro-1-indanone, 98%, ACROS Organics™
Product Code. 10666904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10666904 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10666904 Supplier Thermo Scientific Acros Supplier No. 271120010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 42348-86-7
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00041456
InChI Key MEDSHTHCZIOVPU-UHFFFAOYSA-N
Synonym 5-chloro-1-indanone, 5-chloro-2,3-dihydro-1h-inden-1-one, 5-chloroindan-1-one, 5-chloro-indan-1-one, 1h-inden-1-one, 5-chloro-2,3-dihydro, 5-chloroindanone, 5-chloroindone, pubchem2250, 1-indanone, 5-chloro, 5-chloro indanone
PubChem CID 142599
IUPAC Name 5-chloro-2,3-dihydro-1H-inden-1-one
SMILES ClC1=CC=C2C(=O)CCC2=C1
Specifications
CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 98°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.61
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1-indanone, 98%