missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylcytidine Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05669-100ul
This item is not returnable.
View return policy
Description
5-Methylcytidine Polyclonal antibody specifically detects 5-Methylcytidine in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Methylated DNA Immunoprecipitation
Specifications
| 5-Methylcytidine | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200, Methylated DNA Immunoprecipitation 1:1000-1:2000 | |
| 5 MeCyd, 5 MethylCytidine, 5-MethylCytidine | |
| Chemical compounds corresponding to 5-Methylcytidine/ 5mC. | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Methylated DNA Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction