missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylcytidine Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 516.00
Specifications
| Antigen | 5-Methylcytidine |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:50-1:200, Methylated DNA Immunoprecipitation 1:1000-1:2000 |
| Applications | Immunofluorescence, Methylated DNA Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629906
|
Novus Biologicals
NBP3-05669-100ul |
100 μg |
€ 516.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645008
|
Novus Biologicals
NBP3-05669-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Methylcytidine Polyclonal antibody specifically detects 5-Methylcytidine in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Methylated DNA ImmunoprecipitationSpecifications
| 5-Methylcytidine | |
| Immunofluorescence, Methylated DNA Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 5 MeCyd, 5 MethylCytidine, 5-MethylCytidine | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200, Methylated DNA Immunoprecipitation 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| Chemical compounds corresponding to 5-Methylcytidine/ 5mC. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title