missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Mg, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Aluminum, Al/Mg, 50g
Specifications
Specifications
| Chemical Name or Material | Titanium/Zinc/Gallium/Vanadium/Chromium/Nickel/Lead/Tin/Magnesium/Copper/Iron/Manganese/Silicon/Beryllium |
| CAS | 7429-90-5 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0327 %/Zinc 0.0347 %/Gallium 0.051 %/Vanadium 0.054 %/Chromium 0.0972 %/Nickel 0.142 %/Lead 0.145 %/Tin 0.163 %/Magnesium 0.253 %/Copper 0.287 %/Iron 0.669 %/Manganese 0.752 %/Silicon 0.788 %/Beryllium (<0.0001) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction