Learn More
Stainless Steel, Austenitic, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Stainless Steel, Austenitic, 50g
Specifications
Specifications
| Chemical Name or Material | Tin/Boron/Phosphorus/Cobalt/Aluminium/Nitrogen/Carbon/Copper/Titanium/Vanadium/Silicon/Manganese/Molybdenum/Nickel/Chromium |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tin 0.0097 %/Boron 0.01 %/Phosphorus 0.0149 %/Cobalt 0.0299 %/Aluminium 0.059 %/Nitrogen 0.073 %/Carbon 0.11 %/Copper 0.131 %/Titanium 0.147 %/Vanadium 0.181 %/Silicon 0.44 %/Manganese 0.812 %/Molybdenum 1.05 %/Nickel 5.06 %/Chromium 18.8 % |