missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Austenitic, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Stainless Steel, Austenitic, 50g
Specifications
Specifications
| Chemical Name or Material | Boron/Phosphorus/Vanadium/Nitrogen/Cobalt/Copper/Carbon/Molybdenum/Silicon/Niobium/Manganese/Nickel/Chromium/Tantalum/Aluminium |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0012 %/Phosphorus 0.0409 %/Vanadium 0.0587 %/Nitrogen 0.061 %/Cobalt 0.0702 %/Copper 0.1012 %/Carbon 0.112 %/Molybdenum 0.204 %/Silicon 0.486 %/Niobium 0.487 %/Manganese 0.801 %/Nickel 7.87 %/Chromium 17.45 %/Tantalum (0.012) %/Aluminium (0.03) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction