missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Galvanizing, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, Galvanizing, 50g
Specifications
Specifications
| Chemical Name or Material | Bismuth/Silver/Vanadium/Manganese/Arsenic/Lead/Silicon/Copper/Iron/Aluminium/Magnesium/Zinc |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Bismuth -4 ppm/Silver 0.0003 %/Vanadium 0.0004 %/Manganese 0.0008 %/Arsenic 0.0013 %/Lead 0.003 %/Silicon 0.003 %/Copper 0.009 %/Iron 0.033 %/Aluminium 0.96 %/Magnesium 3.01 %/Zinc (95.96) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction