missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Residuals, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Residuals, 50g
Specifications
Specifications
| Chemical Name or Material | Nickel/Platinum/Selenium/Tellurium/Arsenic/Copper/Tin/Silver/Bismuth/Cadmium/Indium/Antimony |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Nickel 0.002 %/Platinum 0.005 %/Selenium 0.005 %/Tellurium 0.01 %/Arsenic 0.046 %/Copper 0.16 %/Tin 0.19 %/Silver 0.28 %/Bismuth 0.47 %/Cadmium 0.5 %/Indium 0.6 %/Antimony 0.75 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction