missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Residuals, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Residuals, 50g
Specifications
Specifications
| Chemical Name or Material | Indium/Arsenic/Mercury/Selenium/Cadmium/Tellurium/Thallium/Silver/Antimony/Copper/Bismuth/Nickel/Tin/Zinc |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Indium 0.00034 %/Arsenic 0.00038 %/Mercury 0.00045 %/Selenium 0.00048 %/Cadmium 0.00052 %/Tellurium 0.0006 %/Thallium 0.00087 %/Silver 0.00089 %/Antimony 0.00092 %/Copper 0.00095 %/Bismuth 0.00878 %/Nickel (0.0005) %/Tin <0.001 %/Zinc <0.001 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction