missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cu, Alloy K500, N05500, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Nickel, Ni/Cu, Alloy K500, N05500, 50g
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Boron/Phosphorus/Magnesium/Zirconium/Chromium/Carbon/Titanium/Manganese/Iron/Aluminium/Copper/Nickel/Cobalt |
| CAS | 7440-02-0 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.001 %/Boron 0.0015 %/Phosphorus 0.0031 %/Magnesium 0.0098 %/Zirconium 0.0343 %/Chromium 0.073 %/Carbon 0.135 %/Titanium 0.632 %/Manganese 0.634 %/Iron 1.162 %/Aluminium 3 %/Copper 29.91 %/Nickel 64.3 %/Cobalt (0.009) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction