missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, XM-32, M152, S64152, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Stainless Steel, XM-32, M152, S64152, 50g
Specifications
Specifications
| Chemical Name or Material | Tin/Phosphorus/Cobalt/Aluminium/Nitrogen/Copper/Carbon/Silicon/Vanadium/Manganese/Molybdenum/Nickel/Chromium |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tin 0.0053 %/Phosphorus 0.0123 %/Cobalt 0.0185 %/Aluminium 0.0315 %/Nitrogen 0.0339 %/Copper 0.0622 %/Carbon 0.114 %/Silicon 0.224 %/Vanadium 0.275 %/Manganese 0.666 %/Molybdenum 1.567 %/Nickel 2.5 %/Chromium 11.34 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction