missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solder, Lead-Free, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Solder, Lead-Free, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Nickel/Lead/Aluminium/Silver/Germanium/Copper |
| CAS | 7440-31-5 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0086 %/Nickel 0.031 %/Lead 0.0467 %/Aluminium 0.068 %/Silver 0.079 %/Germanium 0.479 %/Copper 0.713 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction