missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al/Cu, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, Zn/Al/Cu, 50g
Specifications
Specifications
| Chemical Name or Material | Iron/Tin/Cadmium/Lead/Bismuth/Manganese/Nickel/Magnesium/Copper/Aluminium/Silicon/Antimony |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Iron 0.0017 %/Tin 0.003 %/Cadmium 0.0033 %/Lead 0.0062 %/Bismuth 0.0113 %/Manganese 0.0153 %/Nickel 0.0258 %/Magnesium 0.048 %/Copper 2.69 %/Aluminium 4.79 %/Silicon (0.0012) %/Antimony (0.0023) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction